Skip to Content
MilliporeSigma
All Photos(1)

Documents

XX2004704

Millipore

Funnel & locking ring assembly, stainless steel

Synonym(s):

Funnel assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

Nylon wheel (locking)
anodized aluminum lock ring
stainless steel funnel(s)

Quality Level

technique(s)

vacuum filtration: suitable

diam.

11.4 cm

funnel capacity

650 mL

application(s)

filtration

shipped in

ambient

General description

The Hydrosol filter holder is designed for the filtration of flammable liquids. Unlike other filter holders, the Hydrosol filter holder includes a grounding unit to prevent sparking from static discharge, which can be a safety hazard.



Applications:

Vacuum Filtration of Flammable Liquids



XX2004720 Hydrosol Filter Holder, 47 mm, stainless steel



Replacement Parts

XX2004704 Funnel and Locking Ring Assembly, stainless steel

XX2004708 Support Screen, 47 mm, stainless steel

XX2004702 Base with Screen, gasket and support

XX2004703 Gasket, PTFE

XX2004718 No. 8 Stopper, 9.5 mm (3/8 in.) hole, silicone

XX20047RK Replacement Parts Kit for Hydrosol Filter Holder



Accessories

XX1004705 Vacuum Filtering Flask, 1 L

XX1004744 Vacuum Filtering Flask, 4L

XX7100004 Tubing 3/16 in. I.D. x 140 cm, silicone

XX6200006P Filter Forceps, blunt end, stainless steel

XX2604735 3-place Manifold, 47 mm, PVC

XX2504735 3-place Manifold, 47 mm, stainless steel

XX2504700 6-place Manifold, 47 mm, stainless steel

WP6211560 High Output Pump, 115 V/60 Hz

WP6222050 High Output Pump, 220 V/50 Hz

WP6210060 High Output Pump, 100 V/50-60 Hz

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service