UFSC400MT
Amicon® Stirred Cell 400mL Maintenance Kit (stir bar assembly, 2 each seals, tubing kit)
Synonym(s):
400mL Stirred Cell Kit, Amicon Maintenance Kit
About This Item
Recommended Products
material
Nylon connector (Luer; on filtrate tubing)
Tygon® tubing (for filtrate; E-3603)
polyethylene tubing (pressure)
polysulfone stir bar(s)
silicone O-ring
silicone gasket
Quality Level
product line
Amicon®
manufacturer/tradename
Amicon®
parameter
400 mL process volume
technique(s)
protein clean-up: suitable (desalting)
protein purification: suitable (concentration)
H
18.7 cm (7.4 in.)
filter diam.
76 mm
filtration area
41.8 cm2
shipped in
ambient
Application
Principle
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.
Protocols
User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service