Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TX1280

Supelco

Trifluoroacetic Acid Soltion

0.1% (v/v) TFA Mobile Phase in Water

Synonym(s):

hplc

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
$543.00
20 L
$2,920.00

$543.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 L
$543.00
20 L
$2,920.00

About This Item

UNSPSC Code:
50202301
grade:
HPLC grade
application(s):
general analytical

$543.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Water/TFA 0.1% Mobile Phase,

Quality Level

grade

HPLC grade

form

liquid

contains

0.095-0.105 % (v/v) trifluoroacetic acid

dilution

(for analytical testing)

evapn. residue

≤5 ppm

color

APHA: ≤10

density

1.000  g/cm3

UV absorption

λ: 205 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.70
λ: 230 nm Amax: ≤0.30
λ: 250 nm Amax: ≤0.05
λ: 280 nm Amax: ≤0.010
λ: 300 nm Amax: ≤0.010
λ: 350 nm Amax: ≤0.010

suitability

suitable for HPLC

application(s)

general analytical

shipped in

ambient

storage temp.

room temp

Application

TX1280P-1 is packaged in poly coated glass bottles

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (trifluoroacetic acid)0.095-0.105% (v/v)Color (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidGradient at 210 nm0.01AU maxGradient at 220 nm0.001AU maxGradient at 280 nm0.001AU maxUV Abs. at 205 nm1.00AU maxUV Abs. at 210 nm0.70AU maxUV Abs. at 230 nm0.30AU maxUV Abs. at 250 nm0.05AU maxUV Abs. at 280 nm0.010AU maxUV Abs. at 300 nm0.010AU maxUV Abs. at 350 nm0.010AU max

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service