Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TX1200

Supelco

Triethylamine

Synonym(s):

Triethylamine, N,N-Diethylethanamine, (Diethylamino)Ethane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$67.20
500 ML
$84.90
2.5 KG
$240.00
20 L
$989.00

$67.20


Available to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
100 ML
$67.20
500 ML
$84.90
2.5 KG
$240.00
20 L
$989.00

About This Item

Linear Formula:
(CH3CH2)3N
CAS Number:
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-469-4
bp:
90 °C/1013 hPa
vapor pressure:
72 hPa ( 20 °C)

$67.20


Available to ship onMarch 31, 2025Details


Request a Bulk Order

vapor pressure

72 hPa ( 20 °C)

Quality Level

200
300

Assay

≥99.5% (GC)

form

liquid

impurities

≤0.1% Water (H2O)

color

clearAPHA: ≤15

pH

12.7 (15 °C, 100 g/L in H2O)

bp

90 °C/1013 hPa

mp

-115 °C

transition temp

flash point -11 °C

solubility

133 g/L

density

0.73 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

N(CC)(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylamine can be used as:
  • An initiator for cracking heptane to yield ethylene and propylene.
  • A catalyst to synthesize polysubstituted dihydrothiophenes via Domino ring-opening/recyclization reactions.
  • A base in the Swern oxidation of primary and secondary alcohols to the corresponding carbonyl compounds.
  • A solvent in the direct one-pot reductive imination of nitroarenes with aldehydes to synthesize imines.

Analysis Note

AppearanceClear, free of particulate matterAssay99.5% minColor (APHA)15 maxIdentity (IR-spectrum)ConformsWater (H2O)0.1% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What grade is TX1200 Triethylamine?

    1 answer
    1. This product is considered a research grade.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service