Skip to Content
MilliporeSigma
All Photos(1)

Documents

TX1056

Supelco

1,2,4-Trichlorobenzene

Meets ACS Specifications OmniSolv®

Synonym(s):

1,2,4-Trichlorobenzene, high purity solvents

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
1,2,4-C6H3Cl3
CAS Number:
MDL number:
UNSPSC Code:
12352101
EC Index Number:
204-428-0

vapor pressure

1.3 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.02% Water (H2O)
≤0.27 μeq/g Titrable acid

color

APHA: ≤10

bp

213.5 °C/1013 hPa

mp

17 °C

transition temp

flash point 99 °C

solubility

0.049 g/L

density

1.45 g/cm3 at 20 °C

UV cutoff

≤308 nm

UV absorption

λ: 308 nm Amax: ≤1.00
λ: 310 nm Amax: ≤0.50
λ: 325 nm Amax: ≤0.10
λ: 375 nm Amax: ≤0.05
λ: 385 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Application

Meets ACS Specifications OmniSolv®
TX1056P-1 is packaged in poly coated glass bottles

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.0% minColor (APHA)10 maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.27µeq/g maxUV Abs. at 308 nm1.00AU maxUV Abs. at 310 nm0.50AU maxUV Abs. at 325 nm0.10AU maxUV Abs. at 375 nm0.05AU maxUV Abs. at 385 nm0.01AU maxUV Cut-off308nm maxWater (H2O)0.02% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service