Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TU-5ML-B

Millipore

Anti-e (clone MS-16/MS-21/MS-63), Blood Grouping Reagent

BIOSCOT Anti-e from the clone MS-16/MS-21/MS-63, Human IgM, Blood Grouping Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

clone

MS-16/MS-21/MS-63, monoclonal

packaging

pkg of 5 mL

manufacturer/tradename

BIOSCOT

storage condition

do not freeze

concentration

15-30 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgM

application(s)

clinical testing

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 30 months from date of filtration

CE Mark

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Labeled

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service