Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TNK1

Millipore

mPAGE® Replacement Tank

replacement part for mPAGE® Mini Gel Tank and mPAGE® Wet Transfer System

Synonym(s):

electrophoresis tank, mini gel tank

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$104.00

$104.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$104.00

About This Item

UNSPSC Code:
41105305
NACRES:
NC.01

$104.00


Please contact Customer Service for Availability

packaging

pkg of 1 pack

manufacturer/tradename

Millipore

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

tank L × W × H

17 cm × 13 cm × 15.5 cm

compatibility

for use with mPAGE® Mini Gel Tank and mPAGE® Mini Wet Transfer System

General description

mPAGE® Replacement Tank for use with mPAGE® Mini Gel Tank and mPAGE® Mini Wet Transfer SystemThe mPAGE® Mini Gel Tank is used to perform high-resolution protein separation on handcast or precast polyacrylamide gels. The gel tank is designed for use with 8 cm x 10 cm mini gels and is compatible with gels cast on the mPAGE® Gel Caster and mPAGE® Precast Gels. The mPAGE® Mini Gel Tank is available in 2 formats for running up to 2 or 4 mini gels and is designed to prevent incorrect assembly and leakage.
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Introduction to PAGE. Learn about SDS-PAGE background and protocol for the separation of proteins based on size in a poly-acrylamide gel.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service