The polycarbonate membranes are translucent when wet, but not considered to be microscopically transparent.
TETP04700
Polycarbonate Membrane Filter, 8.0 μm Pore Size
Isopore™, filter diam. 47 mm, hydrophilic
Synonym(s):
Isopore Membrane Filter
Select a Size
$250.00
Select a Size
About This Item
$250.00
material
plain filter
polycarbonate membrane
white filter
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
Isopore™
Millipore
sustainability
Greener Alternative Product
parameter
140 °C max. temp.
250 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)
60 L/min-cm2-psi air flow rate
~875 mL/min-cm2 water flow rate at 10 psi (typical results)
diam.
47 mm
filter diam.
47 mm
thickness
18 μm
gravimetric extractables
<1%
refractive index
n/D 1.6
matrix
Isopore™
pore size
5-20 % porosity
8.0 μm pore size
greener alternative category
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Application
- Chemotaxis
- Bioassays
- Cytology
- Air monitoring
- Microplastics analysis
Caution
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
-
Hi, are the 'hydrophilic polycarbonate membranes (47 mm diameter)' clear/transparent when wet?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service