Key Documents
TANKLKXL1
Milli-Q
Water sensor
For high volume water purification systems
Select a Size
1 EA
$260.00
$260.00
Please contact Customer Service for Availability
Request more information
Select a Size
Change View
1 EA
$260.00
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
$260.00
Please contact Customer Service for Availability
Request more information
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
This water sensor is a safety accessory which detects water spillage on the floor and automatically closes the system′s inlet solenoid valve to shut down the feed water access from the system or main water supply. The water sensor is typically placed on the floor or benchtop, where most convenient for the lab.
Application
This water sensor accessory for high volume water purification system prevents water damage to the lab, in an unlikely event of a leak.
Features and Benefits
  • Highly reduces the risk of flooding.
  • Protects instruments and lab surroundings from water damage.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany