Documents
SYMBLTVP1
Millipore
Steritest® Symbio Pumps Validation Protoco
Letter format
About This Item
UNSPSC Code:
41116500
NACRES:
JA.11
Quality Level
compatibility
for use with Steritest® Symbio Pumps
General description
Steritest® Symbio Pumps validation protocol is designed to make it easier for the technicians to ensure compliance of the symbio pumps for consistent performance. The product is GMP compliant and includes Validation Master Plan (VMP), Installation Qualification (IQ), Operational Qualification (OQ) and Performance Qualification (PQ) test protocols. In addition to the above, ready to be filled report table formats are also provided.
Application
Steritest® Symbio Pumps validation protocol is a ready to use GMP compliant validation protocol along with ready to be filled report table formats.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany