Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX1075

Sigma-Aldrich

Sucrose

Meets ACS Specifications GR ACS

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 KG
$168.00
12 KG
$334.00
45 KG
$1,140.00

$168.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2.5 KG
$168.00
12 KG
$334.00
45 KG
$1,140.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-334-9

$168.00


Please contact Customer Service for Availability

Request a Bulk Order

form

crystals

color

white

pH

7 (20 °C, 100 g/L in H2O)

mp

185-187 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications GR ACS

Analysis Note

Chloride (Cl)0.005% maxColorWhiteFormCrystalsHeavy metals (as Pb)5ppm maxInsoluble matter0.005% maxInvert sugar0.05% maxIron (Fe)5ppm maxLoss on drying (105°C)0.03% maxResidue after ignition0.01% maxSpecific rotation [a]25/D+66.3-+66.8°Sulfate and sulfite (as SO4)0.005% maxTitrable acid0.0008meq/g max

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service