Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX1045

Supelco

Succinic Acid, Disodium Salt, Hexahydrate

Synonym(s):

Succinic Acid, Disodium Salt, Hexahydrate, Disodium Succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Na2O4 · 6H2O
CAS Number:
Molecular Weight:
270.14
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-778-7
grade:
for analytical purposes
form:
crystals
solubility:
349 g/L

grade

for analytical purposes

Quality Level

Assay

≥98% dry basis

form

crystals

loss

≤40% loss on drying

color

white

pH

6.0-8.5 (20 °C, 50 g/L in H2O)

mp

120 °C (Elimination of water of crystallisation)

solubility

349 g/L

shipped in

ambient

storage temp.

room temp

InChI

1S/C4H6O4.2Na.6H2O/c5-3(6)1-2-4(7)8;;;;;;;;/h1-2H2,(H,5,6)(H,7,8);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

ZBTUYCUNQBRXOR-UHFFFAOYSA-L

Analysis Note

Assay (dried basis): 98% min
Color: White
Form: Crystals
Loss on drying: 40% max

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service