Skip to Content
MilliporeSigma
All Photos(1)

Documents

SX0760

Supelco

Sodium sulfate

anhydrous, crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Disodium Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-820-9

Agency

meets USP/NF

Quality Level

Assay

≥99.0%

form

crystals

impurities

≤0.01% Insoluble matter

loss

≤0.5% loss on ignition

color

white

pH

5.2-8.0 (20 °C, 50 g/L in H2O)

mp

884 °C (lit.)

solubility

200 g/L

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
N: ≤5 ppm
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

anhydrous, crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay99.0% minCalcium (Ca)0.01% maxChloride (Cl)0.001% maxColorWhiteFormCrystalsHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.01% maxIron (Fe)0.001% maxLoss on ignition0.5% maxMagnesium (Mg)0.005% maxNitrogen compounds (as N)5ppm maxPhosphate (PO4)0.001% maxPotassium (K)0.01% maxpH of a 5% solution at 25°C5.2-9.2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service