Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0300

Sigma-Aldrich

Sodium Azide

Synonym(s):

Azium

Sign Into View Organizational & Contract Pricing


About This Item

form

crystals

bp

300 °C/1013 hPa (rigorous decomposition)

mp

275 °C (decomposition)

solubility

408 g/L

storage temp.

room temp

Application

Practical

Analysis Note

Assay:97% min
Color:Off-white to light yellow
Form:Crystals

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service