Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0143V

Supelco

Silica Gel Grade 49

size 6-18 mesh , indicating

Synonym(s):

Silica gel, Amorphous Silicon Dioxide, Amorphous Silica

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
EC Index Number:
231-545-4

form

granular

size

6-18 mesh , indicating

color

blue

particle size

6-18 mesh (>=75%)

bp

2230 °C

mp

>1600 °C

shipped in

ambient

storage temp.

room temp

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-18 Mesh Indicating

Analysis Note

Color: Blue
Form: Granular
Mesh size (6-18): 75% min

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service