Skip to Content
MilliporeSigma
All Photos(1)

Documents

SX0143L

Supelco

Silica Gel Grade 923

100-200 Mesh

Synonym(s):

Silica gel, Amorphous Silicon Dioxide, Amorphous Silica

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
231-545-4

form

powder

size

100-200 mesh

color

white

particle size

100-200 mesh
<2 mm

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

shipped in

ambient

storage temp.

room temp

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

100-200 Mesh

Analysis Note

Color: White
Form: Powder
Mesh size (100 - 200): To pass test

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service