Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SX0075

Supelco

Sand

Standard, washed and dried

Synonym(s):

Sand

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$89.80
2.5 KG
$213.00
12 KG
$607.00

$89.80


Available to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$89.80
2.5 KG
$213.00
12 KG
$607.00

About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
11101503
EC Index Number:
238-878-4

$89.80


Available to ship onMarch 31, 2025Details


Request a Bulk Order

form

granular

impurities

≤0.2% HCl solubles
≤0.2% volatiles

color

beige

particle size

0.2-0.8 mm

pH

5-8 (20 °C, 400 g/L in H2O)

bp

>2200 °C/1013 hPa

mp

1713 °C

anion traces

chloride (Cl-): ≤0.015%

shipped in

ambient

storage temp.

room temp

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Not intended for GC or LC Applications

Analysis Note

Chloride (Cl): 0.015% max
Color: Beige
Description: To pass test
Form: Granules
Soluble substances (in HCl): 0.2% max
Volatile substances: 0.2% max

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service