Skip to Content
MilliporeSigma
All Photos(1)

Documents

ST1698

Sigma-Aldrich

Anti-PDLIM7 Mouse mAb (2D6)

liquid, clone 2D6, Calbiochem®

Synonym(s):

Anti-PD2 and LIM Domain7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D6, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM7(9260)

General description

Purified mouse monoclonal antibody. Recognizes the ~45-50 kDa PDLIM7 protein.
Recognizes the ~45-50 kDa PDLIM7 protein in HeLa cells.
This Anti-PDLIM7 Mouse mAb (2D6) is validated for use in ELISA, Immunoblotting, Immunocytochemistry for the detection of PDLIM7.

Immunogen

Full-length, recombinant, human PDLIM7 (aa 1-458), expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Herrick, S., et al. 2009. Mol. Cell Neurosci.43, 188.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service