Skip to Content
MilliporeSigma
All Photos(1)

Documents

SK1M011W01

Millipore

Pellicon® 2 Cassette Filter

NMWCO 10 kDa

Synonym(s):

Pellicon 2 B10K C 0.1M2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

material

polyethersulfone

Quality Level

quality

400

manufacturer/tradename

Pellicon®

technique(s)

analytical sample preparation: suitable
protein purification: suitable

membrane nominal area

0.1 m2

matrix

Biomax®

pore size

10 kDa NMWCO

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Mini Cassette

Features and Benefits

Type C screen (coarse screen)

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service