Change View
| Size/SKU | Availability | Price |
|---|---|---|
3 each | Please contact Customer Service for Availability | $3,410.00 |
About This Item
UNSPSC Code:
23151806
$3,410.00
Please contact Customer Service for Availability
Product Name
Millipak® 200 CapsuleMedia: Durapore® 5.0 µm, Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF)Inlet: 1-1/2 in. Sanitary FlangeOutlet: 1-1/2 in. Sanitary Flange
Quality Segment
Legal Information
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany