Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCT242

Sigma-Aldrich

BioTracker CDg16 Activated Macrophage Probe

Synonym(s):

Live Cell Imaging Probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

mol wt

calculated mol wt 597.54 kDa

Quality Level

packaging

vial of 1 mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell based assay: suitable
cell culture | stem cell: suitable
flow cytometry: suitable

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C

General description

BioTracker CDg16 activated macrophage probe is specific for activated macrophages and useful for visualizing active inflammatory sites of atherosclerosis in vivo and for targeting the orphan transporter SLC18B1 (Park, 2019).
Reference: Park, Sung-Jin, et al. "Imaging inflammation using an activated macrophage probe with Slc18b1 as the activation-selective gating target." Nature communications 10.1 (2019): 1-7.

Application

Purity > 98% confirmed by UPLC. Identification confirmed by H NMR, F NMR, LC-MS, and elemental analysis.

Features and Benefits

Small molecule green fluorescent probe specific for activated macrophages for imaging inflammation.

Physical form

Lyophilized. Yellow solid.

Storage and Stability

Store at -20°C, desiccated and protected from light. Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Other Notes

Small molecule probe specific for activated macrophages for imaging inflammation.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service