SCT124
GelGreen® Nucleic Acid Stain 10000X DMSO
GelGreen is a fluorescent nucleic acid stain designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels.
Synonym(s):
Fluorescent DNA gel stain
Select a Size
$227.00
Select a Size
About This Item
$227.00
Recommended Products
form
liquid
technique(s)
PCR: suitable
solubility
water: miscible
suitability
suitable for nucleic acid staining
detection method
fluorometric
General description
The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining. GelGreen successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization, under which GelGreen is not classified as hazardous waste.
Spectral Properties
Absorbance: Standard Transilluminator (254 nm) such as a Dark Reader
Emission: Long path green filter such as a SYBR filter or GelStar filter.
Application
Cell Culture
Live Cell Dye
Quality
Absorbance: Standard Transilluminator (254 nm) such as a Dark Reader
Emission: Long path green filter such as a SYBR filter or GelStar filter.
Physical form
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
188.6 °F - (refers to pure substance)
Flash Point(C)
87 °C - (refers to pure substance)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service