SCT066
BioTracker Tau filaments pFTAA Live Cell Dye
Synonym(s):
Live cell imaging probe
Select a Size
$590.00
Select a Size
About This Item
$590.00
Recommended Products
technique(s)
cell based assay: suitable
General description
The BioTracker Tau filaments pFTAA live cell dye is a novel fluorescent probe that can bind to and discriminate between amyloid-b (A?) plaques and tau neurofibrillary tangles (NFTs) in Alzheimer′s disease (AD)-like pathology in transgenic mouse brain tissue sections.
Spectral Properties:
- Excitation: 405 nm
- Emissions: 550 nm
References:
Klingstedt T et al. The Structural Basis for Optimal Performance of Oligothiophene-Based Fluorescent Amyloid Ligands: Conformational Flexibility is Essential for Spectral Assignment of a Diversity of Protein Aggregates. Chem Eur J. 2013, 19, 10179-10192.
Application
Quality
Molar Mass: 704.65 g/mol
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service