Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCT062

Sigma-Aldrich

BioTracker Cys Hcy GSH Triple Live Cell Dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
NACRES:
NA.32

type

for cell culture

concentration

(Please refer to lot specific datasheet.)

technique(s)

cell based assay: suitable

detection method

fluorometric

Application

Live cell fluorescent imaging

Quality

Purity: ≥ 98% confirmed by HNMR, LC-MS and HPLC and elemental analysis
Molar Mass: 435.93 g/mol

Physical properties

435.93 g/mol

Physical form

Lyophilized

Storage and Stability

Store BioTracker Cys Hcy GSH Triple Live Cell Dye at -20°C, desiccate and protect from light Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service