Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCT037

Sigma-Aldrich

BioTracker Far-red Labile Fe2+ Dye

Live Cell imaging probe for labile ferrous (Fe2+) ions, localizing inside endoplasmic reticulum (ER). Also suitable for flow cytometry.

Synonym(s):

ER and golgi Fe2+ probe, cellular iron II ion probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

form

lyophilized (solid blue)

application(s)

cell analysis

detection method

fluorometric

General description

Iron, the most abundant transition metal in our bodies, is involved in a number of biologically important processes such as respiration, oxygen transport, and energy production in collaboration with oxygen. In living cells, iron exists mainly as ferrous (Fe2+) and ferric (Fe3+) ionic forms. As a major contributor to oxidative damage of cells, Fe2+ is implicated in serious diseases such as cancers and neurodegenerative disorders, because of its ability to produce harmful reactive oxygen species via contact with oxygen, superoxide, and hydrogen peroxide (H2O2).BioTracker far-red labile Fe2+ dye is a fluorescent probe that specifically detects labile iron (II) ions (Fe2+). This probe is designed to selectively react with only Fe2+ separately from other metal ions and irreversibly turns into a far-red fluorescent substance. It mainly localizes inside endoplasmic reticulum (ER) and/or golgi bodies. It can also be used with a flow cytometer equipped with a red laser. FerroFarRed shows almost no cytotoxicity at concentrations up to 100 μM (~20 times that of generally used concentrations).This product is same as FerroFarRedTM (GC903-01) from Goryo Chemical, Inc.Spectral Properties:Absorbance (max): 646nmEmission (max): 662nm (Far-red)Research Category:Live Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service