Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCT036

Sigma-Aldrich

BioTracker 625 Red GSH Dye

Live cell imaging dye for glutathione (GSH) that detects reactive oxygen species (ROS) free radicals.

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Glutathione (GSH) is an important antioxidant preventing damage to important cellular components caused by reactive oxygen species (ROS) including free radicals, peroxides, lipid peroxides, and heavy metals. Alterations in glutathione homeostasis are associated with a variety of diseases; therefore, real-time live-cell imaging and quantification of GSH dynamics are important for understanding pathophysiological processes.

The BioTracker 625 Red GSH Dye is probe for glutathione (GSH) in living cells. The dye is formed with an orange fluorescent dye (T) and a deep-red fluorescent dye (S). At a low GSH concentration, green light excitation of T, leads to an energy transfer to S and a subsequent emission of deep-red fluorescent light. At a high GSH concentration, the energy transfer does not occur, so the probe emits the orange fluorescent light from the emission of T. The dye produces a quick and reversible response to the GSH concentration changes, relatively slow photobleaching and low cytotoxicity.

Spectral Properties
Absorbance: 520nm
Emission: 625nm and 582nm

Application

Live cell fluorescent imaging
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Storage and Stability

Store BioTracker 625 Red GSH Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service