Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCR530

Sigma-Aldrich

STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Reprogramming Kit

The STEMCCA Constitutive Polycistronic Lentivirus Kit contains high titer polycistronic lentivirus & Polybrene transfection reagent that have been validated for the generation of mouse & human induced pluripotent stem cells.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.81

Quality Level

manufacturer/tradename

STEMCCA

technique(s)

cell culture | stem cell: suitable

input

sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)

shipped in

dry ice

General description

The STEMCCA Constitutive Polycistronic (OKSM) Lentivirus Kit contains high titer polycistronic (OKSM) lentivirus and Polybrene transfection reagent that have been validated for the generation of mouse and human induced pluripotent stem cells from mouse embryonic fibroblasts (MEFs) and human foreskin fibroblasts (HFFs), respectively. The STEMCCA vector is comprised of the transcription factors Oct-4, Klf4, SOX-2, and c-Myc (OKSM), separated by the self-cleaving 2A peptide and IRES sequences (Sommer CA, 2009). The use of a single polycistronic lentiviral vector significantly improves reprogramming efficiencies and reduces the number of viral integrations.

Components

1. EF1α-STEMCCA (OKSM) Lentivirus: (Part number CS204279) Three (3) vials containing 15 µL of high titer lentivirus.

2. Polybrene Transfection Reagent: (Part number TR-1003-50UL) One (1) vial containing 50 µL of 10 mg/mL stock.

Quality

Tested to confirm the generation of iPS cells from p3 mouse embryonic fibroblasts and p6 human foreskin fibroblasts. Other cell types have not been tested and thus similar results can not be guaranteed.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Stem cell reprogramming protocols to generate human induced pluripotent stem cells (iPSCs) including viral and non-viral RNA based methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service