Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCM134

Sigma-Aldrich

PluriSTEM Freeze Medium

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$183.00

$183.00


Please contact Customer Service for Availability


Select a Size

Change View
50 ML
$183.00

About This Item

UNSPSC Code:
41106200
eCl@ss:
32160801
NACRES:
NA.32

$183.00


Please contact Customer Service for Availability

description

Human Embryonic Stem Cells
Induced Pluripotent Stem Cells

Quality Level

type

for cell culture

form

liquid

technique(s)

cell culture | mammalian: suitable

General description

TM=["PLURISTEM"] Freeze Medium is a defined, serum-free medium for the cryopreservation of human ES and iPS cells. TM=["PLURISTEM"] Freeze Medium contains proprietary small molecule inhibitors, dimethyl sulfoxide (DMSO) and is provided as a complete ready-to-use formulation.

Components

PLURISTEM® Freeze Medium (SCM134)

Storage and Stability

Store at -20°C until expiration date on the product label. Before use, thaw PLURISTEM® Freeze Medium at room temperature (15 - 25°C) or overnight at 2 – 8°C. Do not thaw at 37°C. After thawing, aseptically dispense into working aliquots and store at -20°C. Use aliquots within expiration date as indicated on label. Avoid multiple freeze thaw cycles.

Legal Information

PLURISTEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service