Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCM094

Sigma-Aldrich

Human iPS Reprogramming Boost Supplement II

EMD Millipore’s Human iPS Reprogramming Boost Supplement II contains three proprietary small molecules (TGF-β RI Kinase Inhibitor IV, Sodium Butyrate and PS48) and aids in somatic cell reprogramming by 10-15 fold.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71

Quality Level

form

liquid

input

sample type induced pluripotent stem cell(s)

shipped in

dry ice

General description

EMD Millipore’s Human iPS Reprogramming Boost Supplement II contains three proprietary small molecules (TGF-β RI Kinase Inhibitor IV, Sodium Butyrate and PS48) in amounts sufficient to supplement 350 mL of human ES cell maintenance medium. When used in conjunction with the Human STEMCCA lentivirus reprogramming kits (SCR544, SCR545, and SCR548), the Human iPS Reprogramming Boost Supplement II enhances the efficiency of human iPS colony formation by 10 – 15 fold and shortens the timescale to establish fully reprogrammed colonies by 50%. The colonies formed possess the distinctive flat 2D morphology that are reminiscent of normal human ES cell colonies and can be easily passaged. Fully reprogrammed human iPS colonies that are SSEA-4-positive, TRA-1-60-positive and Hoechst-Dim are readily established in approximately 20 – 25 days.

Application

Research Category
Stem Cell Research

Components

1. TGF-β RI Kinase Inhibitor IV Supplement (1000X): (Part No. CS210445) One (1) vial containing 400 μL of the inhibitor in high quality DMSO.

2.Sodium Butyrate Supplement (1000X): (Part No. CS210446) One (1) vial containing 400 µL of the inhibitor in water.

3. PS48 Supplement (1000X): (Part No. CS210447) One (1) vial containing 400 L of the inhibitor in high quality DMSO.

Storage and Stability

Components of the Human iPS Reprogramming Boost Supplement II are stable for at least 6 months from date of receipt when stored at -20°C. Upon first thaw, centrifuge the vial and gently mix the solution. Aliquot into smaller working volumes and freeze at -20°C or -80°C. Upon addition of the components of the Mouse iPS Reprogramming Boost Supplement II to the media, filter the supplemented media with a 0.22 µM filtration unit. Supplemented media is good for up to two weeks when stored at 2-8°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The Simplicon™ RNA Reprogramming Technology is a next generation reprogramming system that uses a single synthetic, polycistronic self-replicating RNA strand engineered to mimic cellular RNA to generate human iPS cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service