Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCM023

Sigma-Aldrich

Human Mesenchymal-LS Expansion Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
eCl@ss:
32160801
NACRES:
NA.76

type

for cell culture

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

General description

Human Mesenchymal-LS Expansion Medium provides a low serum (2%) cell culture environment for human mesenchymal stem cells including those derived from human adipose and bone marrow tissues and ES/iPS cells. This low serum formulation have been shown to grow cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology.
Human Mesenchymal-LS Expansion Medium provides a low serum (2%) cell culture environment for human mesenchymal stem cells including those derived from human adipose and bone marrow tissues and ES/iPS cells. This low serum formulation have been shown to grow cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. Human Mesenchymal-LS Expansion Medium is specific to human cells and should not be used to culture mouse or rat MSCs. The medium is packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge for added convenience. Human Mesecnhymal-LS Expansion Medium contains no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental result.

Application

Formulated for human mesenchymal stem cell culture.

Components

Human Mesenchymal-LS Basal Medium (SCMM-BM) Human Mesenchymal-LS Supplement Kit (SCMM002-S)

Storage and Stability

Basal Medium (SCMM-BM) should be stored at 2 - 8°C. The special UV protective packaging preotects ENDOGRO® Medium from light damage, however users should take care to protect medium from extended exposure to light. Supplement Kit (SCMM002-S) should be stored at -20°C.

Legal Information

ENDOGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service