Skip to Content
MilliporeSigma
All Photos(2)

Documents

SCC731

Sigma-Aldrich

3dGRO® Colorectal Organoids (POP.112.G2)

Synonym(s):

organoid, PDO, cancer organoid, tumor organoid, 3d cell culture, colorectal cancer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.7

biological source

human

Quality Level

packaging

vial of ≥1500 organoids/vial vials

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

Colorectal cancer accounts for roughly 10% of all cancer cases worldwide with more than half of all patients with CRC developing metastatic disease leading to death. Recently, tissue derived organoids have emerged as a more predictive 3 dimensional cell culture model of disease. 3D organoid cultures conserve the original genetic and phenotypic characteristics of the primary tissue allowing for their application in many research fields included drug development, personalized medicine and potential therapeutics. In vitro cultured tumor organoids have also been shown to predict patient response to chemotherapeutics. PDOs derived from colorectal cancer (CRC organoids) have been used for cell modeling and to investigate the function of cancer related driver gene mutations including APC, TP53, KRAS, BRAF, PIK3CA etc.

We are now offering a comprehensive biobank of highly characterized tissue-derived human gastrointestinal organoids from normal and diseased patients. The intestinal organoid biobank contains over 50 highly characterized intestinal organoids from both normal and diseased patients derived from multiple regions of the digestive systems including small intestine (duodenum, ileum), stomach, rectum and colon.

Cell Line Characteristics
  • Sex: Male
  • Age: 74 Years
  • Organ: Colon (Xenograph from Primary Tumor)
  • Disease: Adenocarcinoma (KRAS Mutation: G12A)

Application

  • Viability: >=1500 viable organoids/vial
  • Organoid Growth: Pass
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
  • Mycoplasma Contamination: Negative
  • STR Profile: Pass

Features and Benefits

Cryopreserved patient derived colorectal cancer organoids (PDOs)

Storage and Stability

Store in liquid nitrogen. The organoids can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Legal Information

3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service