SCC446
OSUMMER.2 Mouse NRAS-Mutant Melanoma Cell Line
Human
Select a Size
$1,390.00
Select a Size
About This Item
$1,390.00
Recommended Products
Product Name
OSUMMER.2 Mouse NRAS-Mutant Melanoma Cell Line,
biological source
human
Quality Level
packaging
vial of ≥1X10⁶ cells/vial mg
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell analysis: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
Application
Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
Cells are verified to be of mouse origin and negative for inter-species contamination from rat, human, Chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
Cells are negative for mycoplasma contamination.
OSUMMER.2 NRAS mouse melanoma cell line was derived from a female TN mouse exposed to UVA and UVB radiation. OSUMMER.2 carries the specific NRAS mutation Q61R and can form tumors in vivo without Matrigel injection. WES revealed high SNV (Single Nucleotide variants) and low CNA (Copy Number Alterations) burden, as well as the presence of UV signature mutations. OSUMMER.2 cells express the melanoma markers PMEL and MelanA and exhibit robust growth in culture.
Source
OSUMMER.2 was derived from melanoma induced in TN (Tyr::CreER) mice exposed to UVA or UVB radiation on postnatal day 3 (1).
References
1. Bowman, R. L., et al. (2021). UVB mutagenesis differs in Nras-and Braf-mutant mouse models of melanoma. Life Science Alliance, 4(9).
Features and Benefits
Storage and Stability
Other Notes
Disclaimer
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service