Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCC290

Sigma-Aldrich

D425 Med Human Medulloblastoma Cell Line

D425 Med human medulloblastoma cell line is representative of a common aggressive form of medulloblastoma and is a well-established disease model.

Synonym(s):

D425 Med Cell Line, Human Medulloblastoma Cells, Medulloblastoma Cell Line

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,590.00

$1,590.00


Please contact Customer Service for Availability


Select a Size

Change View
1 VIAL
$1,590.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$1,590.00


Please contact Customer Service for Availability

application(s)

cell analysis

General description

Medulloblastoma is one of the most common malignant brain tumors in children, with the highest number of diagnoses occurring in patients between 6 and 8 years of age (1). Medulloblastomas are subdivided into four subtypes based on molecular genetic characteristics: WNT, sonic hedgehog (SHH), group 3 and group 4 (1). Of these, group 3 is characterized by amplification of the oncogene MYC, one of the most common gene alterations associated with medulloblastoma.

The D425 Med human medulloblastoma cell line is one of the most highly cited cellular models for medulloblastoma and is classified as a subgroup 3, harboring MYC amplification as well as mutant P53 (2). The D425 Med medulloblastoma cell line was derived from tumor cells of 6-year old patient and grows as suspended or lightly-adherent cell clumps in culture. D425 Med medulloblastoma cells are positive for neurofilament proteins but negative for GFAP (3).

<bold>Source:</bold>
The D425 medulloblastoma cell line was derived from biopsy of a tumor from a 6-year-old male patient (2).

Research Category:
Cancer

Cell Line Origin

Human, Cancer Cells

Packaging

&ge;1X106 cells/vial

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, What is the media for culturing the D425 cells? Thanks, Lakmal

    1 answer
    1. These cells can be cultured in MEM (Richter’s modification) (Thermo Fisher Scientific A1048801) containing 20% FBS (Sigma ES-009-B).

      Please see the data sheet for more information and protocol: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/402/857/scc290-ds.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service