Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCC255

Sigma-Aldrich

WT-1 Mouse Brown Preadipocyte Cell Line

WT-1 mouse brown preadipocyte cell line is a useful model for studying adipogenesis and brown adipocyte biology.

Synonym(s):

Brown Preadipocyte Cells, Mouse Brown Preadipocytes, WT-1 Preadipocyte Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

technique(s)

cell differentiation: suitable

General description

Obesity is a growing worldwide health epidemic that has spurred research into new avenues for treatment and control of metabolic disorders. Brown fat, a specialized adipose tissue characterized by increased energy expenditure, has emerged as an area of interest because manipulation of this tissue has potential to protect against diet-induced obesity by altering the balance of energy intake and consumption (1).

The WT-1 mouse immortalized brown preadipocyte cell line is derived from primary brown preadipocytes. WT-1 preadipocytes can be induced to differentiate into brown adipocytes upon treatment with induction and differentiation factors. WT-1 cells express the brown fat differentiation factor BMP7 and brown fat marker uncoupling protein-1 (UCP1) (3) and are a valuable model for investigation into mitochondrial physiology during adipogenesis. The WT-1 cell line has been used to develop 3D assays for adipose tissue (4) and is well suited for a range of questions in cellular metabolic research.

<bold>Source:</bold>
WT-1 cell line was established from primary brown pre-adipocytes isolated from newborn mouse pups and immortalized with Simian virus 40 (SV40) large T antigen (2).

Research Category:
Stem Cell Research

Cell Line Origin

Mouse, Adipocytes

Packaging

&ge;1X106 cells/vial

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service