Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCC251

Sigma-Aldrich

HL-60 DeltaF508-CF Human Promyelocytic Cell Line

HL-60 DeltaF508-CF human promyelocytic cell line is a valuable tool for drug screening and investigating the role of phagocytic defects in CF disease onset and progression.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

application(s)

cell analysis

General description

Cystic fibrosis (CF) is caused by mutations in the CFTR gene, which encodes a chloride and bicarbonate ion channel (1). Among the approximately 150 known disease-causing mutations in CFTR, deletion of the phenylalanine-508 codon (ΔF508) is the most common and results in protein destabilization, misfolding and ultimately degradation (2). Although the most pronounced lung disease symptoms of CF, including chronic bacterial infection and purulent airway obstruction, are highly associated with neutrophils, the lack of cellular models for CF neutrophils has hindered progress in understanding the significance of these cells for CF pathogenesis (3).The HL-60 DeltaF508-CF human promyelocytic cell line is a derivative of the HL-60 promyelocytic leukemia cell line that has been edited via CRISPR/Cas9 technology for homozygous deletion of CFTR F508 (3). Differentiation of HL-60 DeltaF508-CF cells into neutrophils occurs upon treatment with DMSO. Differentiated HL-60 DeltaF508-CF cells demonstrate normal levels of CD11b staining but substantially reduced CFTR surface expression compared to differentiated HL-60 control cells, in addition to compromised bactericidal activity (3). Source:The HL-60 DeltaF508-CF human promyelocytic cell line was derived from HL-60 promyelocytic leukemia cells transfected with a hCFTR-Cas9-eGFP plasmid. Single-cell clones were sorted for eGFP expression and screened for homozygous deletion of CFTR F508 via nuclease digestion and DNA sequencing (3).

Cell Line Origin

Human, Cancer Cells

Packaging

≥1X106 cells/vial

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service