Skip to Content
MilliporeSigma
All Photos(1)

Documents

SACE-64-5PK

Millipore

CellASIC® Single Cell Isolation Chip

plate size 64 wells, well volume 1-2.5 μL, plate diam. × H 60 mm × 12.6 mm, cell capacity ~20 , single-cell yield

Synonym(s):

Microfluidic single cell isolation chip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

polydimethylsiloxane (PDMS) plate
polystyrene plate

Quality Level

packaging

pk of 5

parameter

100-150 μL sample volume

cell capacity

~20 , single-cell yield

cell size

<50 μm

plate diam. × H

60 mm × 12.6 mm

plate size

64 wells

well diam. × H

1.8 mm × 1 mm

well volume

1-2.5 μL

shipped in

ambient

storage temp.

room temp

General description

The CellASIC® Microfludic single cell isolation chips are microfluidic chips that enable rapid, damage-free isolation of single cells. These devices are less time-consuming and more efficient when compared to limiting dilution (the most common method for single-cell isolation). These devices require no additional instrumentation; they can be loaded with standard pipettes and assayed for single cells using simple microscopy. They require minimal cell numbers and yield single cells in small sample volumes, both attributes are favorable for single cell cloning and genetic/RNA analysis.

Application

Single cell isolation
Whole genome amplification
Target cell isolation
Single Cell PCR

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service