Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S8003

Sigma-Aldrich

CpGenome 5-mC & 5-hmC Human DNA Standards

The CpGenome 5-mC & 5-hmC Human DNA Standards provides two samples of human genomic DNA from the same individual.

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$902.00

$902.00


Please contact Customer Service for Availability


Select a Size

Change View
1 SET
$902.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.31

$902.00


Please contact Customer Service for Availability

Quality Level

species reactivity

human

manufacturer/tradename

CpGenome
Upstate®

shipped in

dry ice

Related Categories

General description

The CpGenome 5-mC & 5-hmC Human DNA Standards provides two samples of human genomic DNA from the same individual. This genomic DNA contains physiological levels of 5-mC and 5-hmC,that have been precisely quantified by LC/MS analysis. These matched human standards are useful as positive controls in DNA methylation assays designed to detect or quantify 5-mC (5-methylcytosine) and/or 5-hmC (5-hydroxymethylcytosine) modifications.

Application

Research Category
Epigenetics & Nuclear Function
The CpGenome 5-mC & 5-hmC Human DNA Standards provides two samples of human genomic DNA from the same individual.

Components

One vial of human brain DNA, 5µg

One vial of human spleen DNA, 5µg

Physical form

Aqueous bufer containing 10 mM Tris-HCl;0.1mM EDTA; pH 8.0.

Storage and Stability

Stable for 12 months at -20°C from date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service