Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7160

Sigma-Aldrich

ApopTag Fluorescein Direct In Situ Apoptosis Detection Kit

Sign Into View Organizational & Contract Pricing

Select a Size

40 ASSAYS
$886.00

$886.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
40 ASSAYS
$886.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

$886.00


Please contact Customer Service for Availability

Request a Bulk Order

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

Quality Level

detection method

fluorometric

General description

Apoptosis is a form of cell death that eliminates compromised or superfluous cells. It is controlled by multiple signaling and effector pathways that mediate active responses to external growth, survival, or death factors. Cell cycle checkpoint controls are linked to apoptotic enzyme cascades, and the integrity of these and other links can be genetically compromised in many diseases, such as cancer. There are many books in print and hundreds of recent review articles about all aspects of apoptosis (e.g. 7, 11, 19, 24, 39, 42) and the methods for detecting it (e.g. 10, 32, 36).

Of all the aspects of apoptosis, the defining characteristic is a complete change in cellular morphology. As observed by electron microscopy, the cell undergoes shrinkage, chromatin margination, membrane blebbing, nuclear condensation and then segmentation, and division into apoptotic bodies which may be phagocytosed (11, 19, 24). The characteristic apoptotic bodies are short-lived and minute, and can resemble other cellular constituents when viewed by brightfield microscopy. DNA fragmentation in apoptotic cells is followed by cell death and removal from the tissue, usually within several hours (7). A rate of tissue regression as rapid as 25% per day can result from apparent apoptosis in only 2-3% of the cells at any one time (6). Thus, the quantitative measurement of an apoptotic index by morphology alone can be difficult.

DNA fragmentation is usually associated with ultrastructural changes in cellular morphology in apoptosis (26, 38).
In a number of well-researched model systems, large fragments of 300 kb and 50 kb are first produced by endonucleolytic degradation of higher-order chromatin structural organization. These large DNA fragments are visible on pulsed-field electrophoresis gels (5, 43, 44). In most models, the activation of Ca2+-and Mg2+-dependent endonuclease activity further shortens the fragments by cleaving the DNA at linker sites between nucleosomes (3). The ultimate DNA fragments are multimers of about 180 bp nucleosomal units. These multimers appear as the familiar âDNA ladderâ seen on standard agarose electrophoresis gels of DNA extracted from many kinds of apoptotic cells (e.g. 3, 7,13, 35, 44).

Another method for examining apoptosis via DNA fragmentation is by the TUNEL assay, (13) which is the basis of ApopTag technology. The DNA strand breaks are detected by enzymatically labeling the free 3′-OH termini with modified nucleotides. These new DNA ends that are generated upon DNA fragmentation are typically localized in morphologically identifiable nuclei and apoptotic bodies. In contrast, normal or proliferative nuclei, which have relatively insignificant numbers of DNA 3′-OH ends, usually do not stain with the kit. ApopTag Kits detect single-stranded (25) and double-stranded breaks associated with apoptosis. Drug-induced DNA damage is not identified by the TUNEL assay unless it is coupled to the apoptotic response (8). In addition, this technique can detect early-stage apoptosis in systems where chromatin condensation has begun and strand breaks are fewer, even before the nucleus undergoes major morphological changes (4, 8).

Apoptosis is distinct from accidental cell death (necrosis). Numerous morphological and biochemical differences that distinguish apoptotic from necrotic cell death are summarized in the following table (adapted with permission from reference 39). ApopTag In Situ Apoptosis Detection Kits distinguish apoptosis from necrosis by specifically detecting DNA cleavage and chromatin condensation associated with apoptosis. However, there may be some instances where cells exhibiting necrotic morphology may stain lightly (14, 29) or, in rare instances, DNA fragmentation can be absent or incomplete in induced apoptosis (11).
It is, therefore, important to evaluate ApopTag staining results in conjunction with morphological criteria. Visualization of positive ApopTag results should reveal focal in situ staining inside early apoptotic nuclei and apoptotic bodies. This positive staining directly correlates with the more typical biochemical and morphological aspects of apoptosis.

Since an understanding of cell morphology is critical for data interpretation and because of the potential for experimentally modifying or overcoming normal apoptotic controls, the following strategy is advised. When researching a new system, the staging and correlation of apoptotic morphology and DNA fragmentation should be characterized. In some tissues, cytoplasmic shrinkage may be indicated by a clear space surrounding the cell. The nuclear morphology of positive cells should be carefully observed at high magnification (400x-1000x). Early staged positive, round nuclei may have observable chromatin margination. Condensed nuclei of middle stages, and apoptotic bodies, usually are stained. Apoptotic bodies may be found either in the extracellular space or inside of phagocytic cells. It is highly recommended that less experienced observers should refer to illustrations of dying cells for comparison with new data (e.g. 11, 19, 24).

An additional, although far less sensitive, method of confirming ApopTag staining results is the detection of DNA fragmentation on agarose gels. If a large percent of the cells in the tissue are apoptotic, then electrophoresis of extracted total genomic DNA and standard dye staining can be used to corroborate the in situ staining. However, the single-cell sensitivity of ApopTag histochemistry is far higher than this method. DNA laddering data of comparable sensitivity may be obtained in several other ways. These include methods for selectively extracting the low molecular weight DNA (15), for preparing radiolabeled DNA (30, 40) in combination with resin-bed purification of DNA (12), and for DNA amplification by PCR (35).

The in situ staining of DNA strand breaks detected by the TUNEL assay and subsequent visualization by light microscopy gives biologically significant data about apoptotic cells which may be a small percentage of the total population (13, 16). Apoptotic cells stained positive with ApopTag Kits are easier to detect and their identification is more certain, as compared to the examination of simply histochemically stained tissues. Another feature of ApopTag is that quantitative results can be obtained using flow cytometry, since end-labeling methodology detects apoptotic cells with a >10-fold higher sensitivity than necrotic cells (14,17). In addition, the occurrence of DNA fragmentation with regard to the cell cycle phase of apoptotic cells can be examined using the TUNEL assay and flow cytometry (16,18).
The ApopTag Fluorescein Direct In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the direct TUNEL method. The DNA fragments are tagged with a directly labeled fluorescein nucleotide. The kit provides fluorescence staining for 40 samples which are analyzed using flow cytometry.

This kit is designed for use on adherent and suspended cells. For tissue sections use our ApopTag kits Catalog Number S7110 or S7111.

In some instances, certain tissues contain cell types which bind any fluorescein labeled nucleotide such as that contained in the ApopTag Fluorescein Direct Kit (S7160). It is therefore recommended that this kit be used to stain cell suspensions, cytospins and cell cultures, but not tissue sections. Chemicon suggests the use of the ApopTag Fluorescein Kits (S7110, S7111) or the ApopTag Red Kit (S7165) for fluorescent staining tissue sections.

Application

INTRODUCTION

ApopTag In Situ Apoptosis Detection Kits label apoptotic cells in research samples by modifying fragmented genomic DNA utilizing terminal deoxynucleotidyl transferase (TdT) for detection of positive cells by specific staining. This manual contains information and protocols for the ApopTag Fluorescein Direct In Situ Apoptosis Detection Kit (Catalog number S7160).

Principles of the Procedure

The reagents provided in ApopTag Kits are designed to label the free 3′OH DNA termini in situ with chemically labeled and unlabeled nucleotides. The nucleotides contained in the Reaction Buffer are enzymatically added to the DNA by terminal deoxynucleotidyl transferase (TdT) (13, 31). TdT catalyzes a template-independent addition of nucleotide triphosphates to the 3′-OH ends of double-stranded or single-stranded DNA. In the ApopTag Direct Methodology, the incorporated nucleotides form an oligomer composed of fluorescein nucleotide and unlabeled nucleotide in a random sequence. The ratio of labeled to unlabeled nucleotide in the kit is optimized to minimize fluorescein self-quenching.
The exact length of the oligomer added has not been measured.The ApopTag Fluorescein Direct In Situ Apoptosis Detection Kit (S7160) does not require the use of an antibody for detection, since the DNA fragments are directly labeled with a fluorescein nucleotide. Because the anti-digoxigenin antibody is not used, this system requires fewer applications of solutions (Figure 1B), which promotes good cell recovery due to fewer manipulations. The protocol also requires less time. There is little difference in the mean signal strength relative to the indirect method (32).

Results using ApopTag Kits have been widely published (see Sec. V. References, Publications Citing ApopTag Kits). The ApopTag product line provides various options in experimental design. A researcher can choose to detect staining by brightfield or fluorescence microscopy or by flow cytometry, depending on available expertise and equipment. There are also opportunities to study other proteins of interest in the context of apoptosis when using ApopTag Kits. By using antibodies conjugated with an enzyme other than peroxidase and an appropriate choice of substrate, it is possible to simultaneously examine another protein and apoptosis using ApopTag Peroxidase Kits. There is also a choice of fluorophores (fluorescein and rhodamine) using ApopTag technology. Flexibility exists in choosing antibody-fluor combinations to study other important proteins.

Components

Equilibration Buffer 90416 3.0 mL -15°C to -25°C Reaction Buffer 90427 2.0 mL -15°C to -25°C TdT Enzyme 90418 0.64 mL -15°C to -25°C Stop/Wash Buffer 90419 20 mL -15°C to -25°C Plastic Coverslips 90421 100 ea. Room Temp. Number of tests per kit: Sufficient materials are provided to stain 40 tissue specimens of approximately 5 cm2 each when used according to instructions. Reaction Buffer will be fully consumed before other reagents when kits are used for slide-mounted specimens.

Storage and Stability

1. Store the kit at -15°C to -25°C until the first use. After the first use, if the kit will be used within three months, store the TdT Enzyme (90418) at -15°C to -25°C and store the remaining components at 2°C to 8°C. 2. Protect the fluorescein nucleotide (90427) in the Reaction Buffer from unnecessary exposure to light. Precautions 1. The following kit components contain potassium cacodylate (dimethylarsinic acid) as a buffer: Equilibration Buffer (90416), Reaction Buffer (90427), and TdT Enzyme (90418). These components are harmful if swallowed; avoid contact with skin and eyes (wear gloves, glasses) and wash areas of contact immediately. 2. TdT Enzyme (90418) contains glycerol and will not freeze at -20°C. For maximum shelf life, do not warm this reagent to room temp. before dispensing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service