Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RX0090

Sigma-Aldrich

Rhodamine 6 G

Used as a dye and a fluorescent probe

Synonym(s):

Rhodamine 6 G, Basic Red 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45160
MDL number:
UNSPSC Code:
41116121
EC Index Number:
213-584-9

form

solid

mp

215 °C

λmax

525-530 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H

InChI key

VYXSBFYARXAAKO-UHFFFAOYSA-N

Analysis Note

Color: Red
Extinction coefficient: 69900 min
Form: Powder
Loss on drying: 5% max
Wavelength of max. absorbance: 525-530nm
pH of a 1% aqueous solution:

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service