Skip to Content
MilliporeSigma
All Photos(1)

Documents

R03379

Sigma-Aldrich

Formalin

ready-to-use solution, Harleco®, 10%

Synonym(s):

Fixative

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352501

form

ready-to-use solution

Quality Level

manufacturer/tradename

Harleco®

concentration

10%

color

colorless

pH

7

mp

-9.21 °C

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO®Ready-to-use fixing solution, 10% Carbonate Buffered

Application

HARLECO®Ready-to-use fixing solution, 10% Carbonate Buffered

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service