Skip to Content
MilliporeSigma
All Photos(1)

Documents

QTUMMPKIX

Milli-Q

Kit: Quantum® IX Cartridge & Millipak®

Kit with Millipak® 40 Express Filter, For use with Milli-Q® water purification systems.

Synonym(s):

Kit: Quantum® IX Cartridge & Millipak® 40 Express Final Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
NACRES:
JA.13

material

PVDF filter
polypropylene and polyethylene cartridge

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Millipak®
Quantum®

compatibility

for use with Milli-Q® Biocel
for use with Milli-Q® Element
for use with Milli-Q® Gradient
for use with Milli-Q® Integral
for use with Milli-Q® Synthesis

Looking for similar products? Visit Product Comparison Guide

General description

This kit is composed of a Quantum® IX Cartridge and a Millipak® 40 Final Filter.
The Quantum® IX Polishing Cartridge contains a combination of purification media carefully selected to remove trace contaminants affecting specific applications.
The cartridge is constructed from polypropylene and polyethylene, which are suitable for the production of ultrapure water and are qualified for low organics and ionic extractables.
The Millipak® 40 filter produces particulate- and bacteria-free water to meet the stringent requirements of scientific applications.

Application

The kit contains Organex and Ionex resins for the removal of trace levels of organics and ions.

Features and Benefits

  • Quantum® IX Polishing Cartridge has been designed to remove trace levels of organics and ions from ultrapure water.
  • Millipak® 40 filter is made with Durapore® PVDF membranes, which provide high flow rates and throughputs, low extractable and broad chemical compatibility.
  • The membrane is supported by a rigid plastic disc with channels that ensures high water flow and resistance to pressure variations without the risk of membrane rupture.
  • Millipak® membrane filters use a unique, clean stacked disc design, allowing minimal hold-up volume and particle shedding.
  • The Millipak® 40 filter connects to the outlet of the water delivery point, such as the PODs of Milli-Q® water purification systems.

Other Notes

Directions for Use

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: General laboratory analysis
  • Intended Use: Water purification
  • Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes. Refer to the system equipment user guide section "Using the System" (provided with the system)
  • Storage Statement: Store in a dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
QUANTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service