Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1824

Supelco

n-Propyl Alcohol

OmniSolv®

Synonym(s):

n-Propyl Alcohol, 1-Propanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$196.00
4 L
$488.00

$196.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$196.00
4 L
$488.00

About This Item

Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-746-9
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
96.5-98 °C/1013 hPa
vapor pressure:
19 hPa ( 20 °C)

$196.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

vapor pressure

19 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99.5% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Aldehydes
≤0.05% Water (H2O)
≤0.2 μeq/g Titrable acid
≤200 ppt Fluorescence (as quinine base)

evapn. residue

≤1 ppm

color

APHA: ≤10

pH

7 (20 °C, 200 g/L in H2O)

bp

96.5-98 °C/1013 hPa

mp

-127 °C

transition temp

flash point 15 °C

density

0.8 g/cm3 at 20 °C

UV cutoff

≤210 nm

UV absorption

λ: 210 nm Amax: ≤1.00
λ: 220 nm Amax: ≤0.50
λ: 230 nm Amax: ≤0.25
λ: 250 nm Amax: ≤0.05
λ: 300 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

OCCC

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Application

OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Aldehydes0.01% maxAssay (GC)99.5% minColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)200ppt maxFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.2µeq/g maxUV Abs. at 210 nm1.00AU maxUV Abs. at 220 nm0.50AU maxUV Abs. at 230 nm0.25AU maxUV Abs. at 250 nm0.05AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off210nm maxWater (H2O)0.05% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service