Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1690

Supelco

1,2-Propanediol

Synonym(s):

1,2-Propanediol, Propylene Glycol, 1

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
$838.00
18.9 L
$2,090.00

$838.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 L
$838.00
18.9 L
$2,090.00

About This Item

Empirical Formula (Hill Notation):
C3H8O2
CAS Number:
Molecular Weight:
76.09
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-338-0
bp:
188 °C/1013 hPa
vapor pressure:
0.11 hPa ( 20 °C)

$838.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.11 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

color

APHA: ≤10

refractive index

n20/D 1.43

pH

6-8 (20 °C, 100 g/L in H2O)

bp

188 °C/1013 hPa

mp

-59 °C

transition temp

flash point 99 °C

density

1.04 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OC(CO)C

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Analysis Note

Assay (GC): 98% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service