Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PX1520

Supelco

Potassium Nitrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Potassium Nitrate, Nitrate of Potash

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$168.00
12 KG
$1,800.00

$168.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$168.00
12 KG
$1,800.00

About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-818-8
grade:
ACS reagent
form:
crystals

$168.00


Estimated to ship onMarch 24, 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

meets USP/NF

Assay

≥99.0%

form

crystals

impurities

≤0.005% Insoluble matter

color

white

pH

5.0-7.5 (20 °C, 50 g/L in H2O)

mp

334 °C

anion traces

chloride (Cl-): ≤0.002%
iodate (IO3-): ≤5 ppm
nitrite (NO2-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤3 ppm
Mg: ≤0.002%
Na: ≤0.005%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[K+].[N+](=O)([O-])[O-]

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay99.0% minCalcium (Ca)0.005% maxChloride (Cl)0.002% maxColorWhiteFormCrystalsHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.005% maxIodate (IO3)5ppm maxIron (Fe)3ppm maxMagnesium (Mg)0.002% maxNitrite (NO2)0.001% maxPhosphate (PO4)5ppm maxSodium (Na)0.005% maxSulfate (SO4)0.003% maxpH of a 5% solution at 25°C4.5-8.5

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service