Skip to Content
MilliporeSigma
All Photos(2)

Documents

PR0G0TNP2

Milli-Q

Progard® Pretreatment Pack

Protects the reverse osmosis (RO) membrane from water components

Synonym(s):

Progard® TNP2 Pretreatment Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

material

EPDM rubber
polypropylene

Quality Level

feature

Optimal protection of Reverse Osmosis (RO) membrane, optimized system lifetime and performance. Removal of large particles, organic compounds and free chlorine from feed water with a silt density index (SDI) up to 12

packaging

pkg of 1 unit

manufacturer/tradename

Progard®

H

39.5 cm (15.5 in.)

total organic carbon (TOC) residue

<2000 ppb

halogenated residue

<3 ppm (Cl2)

compatibility

for use with AFS®
for use with Elix® Advantage
for use with Elix® Essential UV
for use with Elix® Essential
for use with Elix® Reference
for use with Milli-Q® Integral
for use with RiOs Essential

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Progard® TNP2 Pretreatment Pack pretreats tap water by removing large particles, organic compounds and free chlorine. This protects the reverse osmosis (RO) membrane against organic fouling and chlorine oxidation.
Progard® TNP2 Pretreatment Pack ensures that your system lifetime and performance are optimized.
Replacement of consumables, when requested by the system, is key to maintain the system′s performance within specifications.
Designed for use with AFS® Essential, Elix® Advantage, Elix® Reference, Elix® Essential, Milli-Q® Integral, and RiOs Essential water purification systems.

Application

Removes free chlorine, organics and particles to protect the RO membrane.

Features and Benefits

  • The Progard® TNP2 pretreatment pack is constructed from polypropylene and EPDM.
  • Suitable for tap water feed with silt density index (SDI) < 12 and Cl2 < 3 ppm, and TOC < 2000 ppb.
  • Natural activated carbon media exhibits a high surface area and adsorption kinetic, tolerating feed water containing up to 3 ppm of chlorine and 2000 ppb of TOC. Removes free chlorine and organic compounds.
  • To prevent the bacteria proliferation, the natural activated carbon selected has been impregnated with silver, a proven bacteriostatic agent.
  • Depth filter media exhibits a high capacity and optimized retention, tolerating feed water SDI up to 12 and removes particles.
  • 100% traceability with e-Sure Tag - Incorporates RFID technology. This provides instant visibility on pack parameters and ensures a safe and optimal performance of the system, giving full traceability.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service