Skip to Content
MilliporeSigma
All Photos(1)

Documents

PMEF-HL

Sigma-Aldrich

EmbryoMax® Primary Mouse Embryonic Fibroblasts

Mouse

Synonym(s):

Hygro MEF Feeder Cells, Hygro Mouse Feeder Cells, Hygro-MEFs, Hygromycin Restistant PMEFs, MEF Feeder Cells, hygro PMEFs, hygro MEFs, MEFs, Mouse Feeder Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.75

product name

EmbryoMax® Primary Mouse Embryonic Fibroblasts, PMEF, Hygro Resistant, Strain C57/BL6, Not Treated, Passage 3

biological source

mouse

Quality Level

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)
sample type primary embryotic fibroblasts (PMEFs)
sample type induced pluripotent stem cell(s)

shipped in

liquid nitrogen

General description

Primary Mouse Embryo Fibroblasts, hygromycin resistant, strain C57/BL6, are not Mytomycin C treated and serve as a feeder layer for both mouse and human ES/IPS cells. MEF cells are resistant to hygromycin. The cells are provided at passage 3 and can be further expanded before freezing for later use.

Plating MEF Feeder Cells

Procedure:

1. Prior to thawing PMEF feeder cells, coat plates/flasks with Gelatin solution.
2. Thaw PMEF vial(s) quickly in a 37 °C water bath and transfer to a 15 mL tube (already containing 10 mL of warm PMEF Feeder Cell Medium). Gently invert the tube to distribute, and centrifuge at 300 xg for 4–5 minutes.
3. Remove supernatant and resuspend the cell pellet in warm PMEF Feeder Cell Medium.
4. Remove the Gelatin solution from plates/flasks, and aliquot the PMEF feeder cell suspension at the densities recommended in Table 4.1 of the mouse ES protocol guide
5. Incubate the PMEF Feeder cells at 37 °C with 5% CO2. Use Figures 4A, B and C in the mouse ES protocol guide as a guide for an estimate of correct PMEF density and
appearance. Gelatinized plates may be used for 12–14 days.
The EmbryoMax range of PMEF cells provides researchers with a convenient solution for ES cell culture by eliminating the need for time consuming feeder cell isolation and preparation. Many embryonic stem cell culture protocols necessitate the use of primary mouse embryo fibroblast (PMEF) cells. In these protocols, ES cells are typically cultured on a monolayer of PMEF feeder cells. Feeder cells perform two important roles in stem cell culture: they secrete several important growth factors into the medium, which help maintain pluripotency, and they provide a cellular matrix for ES cells to grow.

Cell Line Description

Primary Mouse Embryo Fibroblasts

Packaging

5-6x106 ea

Physical form

The cells are provided frozen into individual vials, containing approximately 5-6 x 106 fibroblasts per vial, and are sold as a pack of 5 vials and supplied at passage 3.

Storage and Stability

After receipt, vials should be stored at -80°C. If storing for longer than 6 months, store in the vapor phase of liquid nitrogen (make sure the cap is tight) for up to 2 years.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

Protocols

Stem Cell protocols for cryopreservation, thawing of cryopreserved stem cells and media preparation.

Stem cell reprogramming protocols to generate human induced pluripotent stem cells (iPSCs) including viral and non-viral RNA based methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service