Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PF138

Sigma-Aldrich

Resistin, Human, Recombinant, E. coli

Synonym(s):

Resistance to Insulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥98% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤100 ng/μg Endotoxin

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human resistin expressed in E. coli. A dimeric protein secreted by adipocytes. Resistin may be an important link between obesity and type II diabetes; levels of resistin have been shown to be altered in animal models of diabetes. Contains ≤0.1 ng endotoxin per µg resistin.
Recombinant, human resistin expressed in E. coli. Resistin is a ~19.5 kDa dimeric protein which is secreted by adipocytes. Resistin may be an important link between obesity and type II diabetes; levels of resistin have been shown to be altered in animal models of diabetes.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 10 mM sodium citrate, pH 3.0.

Reconstitution

Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 week at 4°C or for up to 6 months at -20°C.
Reconstitute with water to a concentration of 0.1-1 mg/ml.

Other Notes

Endotoxin: ≤ 0.1 ng endotoxin/µg resistin
McTernan, P.G., et al. 2002. J. Clin. Endocrinol. Metab.87, 2407.
Ravussin E., and Smith, S.R. 2002. Ann. N.Y. Acad. Sci.967, 363.
Steppan, C.M., et al. 2001. Nature409, 307.
Way, J.M., et al. 2001. J. Biol. Chem.276, 25651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service