Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PCC300C45L

Millipore

Pellicon® Capsule with Ultracel® Membrane

C Screen, 4.5 m2 manifold AseptiQuik® L connectors

Synonym(s):

Pellicon® Capsule with Ultracel® Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806

material

EPDM rubber seal
PPO/PS blend (core
core)

PPO/PS blend housing
TPE seal
composite regenerated cellulose (CRC) membrane
epoxy adhesive
polyester screen
polypropylene seal
polyurethane adhesive

Quality Level

sterility

sterile; γ-irradiated

product line

Pellicon®

feature

holdup volume 1685 mL (in feed channel)
holdup volume 2559 mL (in permeate channel)

packaging

package of 1

parameter

max. inlet pressure at 60 psi (4.1 bar) and 4-30 °C
max. transmembrane pressure at 45 psi (3.1 bar) and 4-30 °C
4-6 L/min-m2 flow rate

technique(s)

ultrafiltration: suitable

filtration area

4.5 m2

screen size

, Type C screen (coarse screen)

size

46.9 cm (18.5 in.) , Height
56.2 cm (22.1 in.) , Length

width

48.2 cm (19 in.)
482 mm

impurities

≤5 ppm Meets TOC (in retentate effluent, after a WFI flush of 20 L/m²)

matrix

Ultracel®

pore size

300 kDa

pH range

2-13

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The Pellicon® Capsule is the first of its kind -- a true single-use TFF device that comes ready to process in minutes. Self-contained, it employs an easy-to-use, holderless format and is supplied sterilized by irradiation.
Feed: AseptiQuik® L Connector
Permeate: AseptiQuik® L Connector
Retentate: AseptiQuik® L Connector
Permeate: AseptiQuik® G Connector

Application

Viral Gene Therapies, Vaccines, Concentration, Diafiltration, Buffer exchange, Protein purification, mRNA

Features and Benefits

  • Plug ′n play, holderless design - easy to install, safe to remove
  • Gamma Sterilized and preseravtive-free - ready to process in minutes
  • True single-use, self-contained capusle - fast, safe, and flexible batch turnaround
  • Proven Ultracel® membrane and C screen - high recovery, superior mass transfer, solvent resistance
  • Pellicon® TFF proven performance - true linear scalability within Pellicon® TFF families

Packaging

The manifold assembly is inserted into a polyethylene foam frame and then double bagged and vacuum sealed. The double-bagged assembly is placed within a second foam frame and individually boxed (qty 1 per box). Packaged assemblies are palletized.

Other Notes

The Emprove® Program complements the product portfolio through

  • Comprehensive documentation to support qualification, risk assessment and process optimization needs.
  • Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
  • Convenient 24/7 access to up-to-date product information.

Legal Information

AseptiQuik is a registered trademark of Colder Products Company
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our overview of scalable tangential flow filtration for viral vector production will provide key considerations for developing and optimizing your downstream process.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service