Skip to Content
MilliporeSigma

P3B010A01

Millipore

Pellicon® 3 Cassette with Biomax® Membrane

10kDa, A Screen, 0.11 m2

Synonym(s):

Pellicon® 3 Cassette with Biomax 10 kDa Membrane, A screen, 0.11 m2

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,890.00

$1,890.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$1,890.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031623

$1,890.00


Please contact Customer Service for Availability

material

TPE seal
polyethersulfone
polypropylene cap (for end)
polypropylene screen

Quality Level

feature

holdup volume 9 mL (in feed channel)

manufacturer/tradename

Pellicon®

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours)
4-6 L/min-m2 recirculation rate
40 psi max. transmembrane pressure at 4-50 °C
6.9 bar max. inlet pressure (100 psi)
80 psi max. transmembrane pressure at 4-40 °C

technique(s)

analytical sample preparation: suitable
buffer exchange: suitable
protein purification: suitable (concentration)

H

2.3 cm (0.90 in.)

L

20.6 cm (8.1 in.)

W

5.6 cm (2.2 in.)

membrane area

0.11 m2

gravimetric extractables

≤1500 mg/m2

matrix

Biomax® (Membrane)

pore size

10 kDa NMWCO

pH

1-14

Looking for similar products? Visit Product Comparison Guide

General description

Pellicon® 3 filtration devices are advanced, high-performance cassettes ideal for today’s higher titer therapeutic antibodies as well as the more demanding filtration processes that require higher operating pressures, temperatures and caustic cleaning regimes.

The Pellicon® 3 Cassette with Biomax® Membrane is perfect for the filtration of monoclonal antibodies and solutions containing albumin, hormones, vaccines and growth factors.

This P3B010A01 Pellicon® 3 ultrafiltration device features a pore size of 10 kDa NMWCO, a Biomax® 10 membrane, and dimensions measuring 2.3 cm (H) x 5.6 cm (W) x 20.6 cm (L).

Features and Benefits

  • Rugged & reliable – ideally suited for today′s more demanding filtration processes
  • High performance – produces excellent resolution, yields and back-pressure resistance
  • Optimum product recovery – thanks to proven membrane technology
  • Void-free technology – leading-edge membranes resolve virtually any separation challenge
  • Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
  • Automated manufacturing – delivers unbeatable performance, consistency and reliability
  • Easy installation & care – simple to install and maintain

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Analysis Note

Gravimetric Extractables: after 24 hours in water at controlled room temperature

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service