Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2PLACV05

Millipore

Pellicon 2 Cassette Ultrafiltration Module PLAC V 0.5 m2

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$5,653.00

$5,653.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$5,653.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031623

$5,653.00


Please contact Customer Service for Availability

material

polypropylene screen
polyurethane
regenerated cellulose

Quality Level

description

Filter code PLAC
Type V screen (open channel)
prefiltration requirements: 100 µm

parameter

3.5 bar max. transmembrane pressure (50 psig) at 30 °C
5-35 L/min recirculation rate at 0.4 bar (6 psi)

technique(s)

analytical sample preparation: suitable
protein purification: suitable
tangential flow filtration (TFF): suitable
ultrafiltration: suitable

H

2.16 cm (0.85 in.)

L

21 cm (8.3 in.)

device size

0.5 m2

filtration area

0.5 m2

membrane nominal area

0.1 m2

width

17.8 cm , 7.0 in.

matrix

Ultracel®

pore size

1 kDa filter

Analysis Note

The extractables level was equal to or less than 1500 mg/m² after a water flush.

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service